Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite 96%
Leverantör: Apollo Scientific
Varning
Synonyms: Bis(2-cyanoethyl) diisopropylphosphoramidite, Bis(2-cyanoethoxy)(diisopropylamino)phosphine, Bis(2-cyanoethoxy)-N,N-diisopropylaminophosphine
OR4200T-1G OR4200T-5G OR4200T-10G
APOSOR4200T-1GEA 1200 SEK
APOSOR4200T-1G APOSOR4200T-5G APOSOR4200T-10G
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite 96%
Bis(2-cyanoethyl)-N,N-diisopropylphosphoramidite
A useful phosphorylating reagent.
Formel: C₁₂H₂₂N₃O₂P
MW: 271,301 g/mol
Storage Temperature: Frys
MDL Number: MFCD00797597
CAS nummer: 102690-88-0
UN: 3278
ADR: 6.1,III