N-(Ethoxycarbonyl)nortropinone 97%

Leverantör: Apollo Scientific
Varning

Synonyms: Ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate, 8-(Ethoxycarbonyl)-8-azabicyclo[3.2.1]octan-3-one, N-Carbethoxy-4-tropinone

OR41153-25G OR41153-10G
APOSOR41153-25GEA 1525 SEK
APOSOR41153-25G APOSOR41153-10G
N-(Ethoxycarbonyl)nortropinone 97%
N-(Ethoxycarbonyl)nortropinone
Formel: C₁₀H₁₅NO₃
MW: 197,23 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00078171
CAS nummer: 32499-64-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR