Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid 97%
Leverantör: Apollo Scientific
Varning
OR41128-10G OR41128-25G OR41128-5G
APOSOR41128-10GEA 1300 SEK
APOSOR41128-10G APOSOR41128-25G APOSOR41128-5G
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid 97%
Pyrazolo[1,5-a]pyrimidine-3-carboxylic acid
Formel: C₇H₅N₃O₂
Storage Temperature: Omgivande
MDL Number: MFCD01975194
CAS nummer: 25940-35-6