Isothiazolin-3-one
Leverantör: Apollo Scientific
Varning
Synonyms: Isothiazol-3(2H)-one
OR40800-1G OR40800-250MG
APOSOR40800-1GEA 755 SEK
APOSOR40800-1G APOSOR40800-250MG
Isothiazolin-3-one
Isothiazolin-3-one
Formel: C₃H₃NOS
MW: 101.13 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09834764
CAS nummer: 1003-07-2