Acrolein diethyl acetal 98%
Leverantör: Apollo Scientific
Fara
Synonyms: Acrolein Acetal, 1,1-Diethoxy-2-propene, 3,3-Diethoxy-1-propene
OR40659-100G OR40659-250G OR40659-25G
APOSOR40659-100GEA 940 SEK
APOSOR40659-100G APOSOR40659-250G APOSOR40659-25G
Acrolein diethyl acetal 98%
Acrolein diethyl acetal
Formel: C₇H₁₄O₂
Kokpunkt: 120…125 °C
Densitet: 0,852 g/cm³ (25 °C)
Flampunkt: 13 °C
Storage Temperature: Omgivande
MDL Number: MFCD00009239
CAS nummer: 3054-95-3
UN: 2374
ADR: 3,II