Ethyl-2-amino-4-methyl-5-oxazolecarboxylate 95+%

Leverantör: Apollo Scientific
Varning

Synonyms: Ethyl 2-amino-4-methyloxazole-5-carboxylate

OR40536-1G OR40536-500MG
APOSOR40536-1GEA 1050 SEK
APOSOR40536-1G APOSOR40536-500MG
Ethyl-2-amino-4-methyl-5-oxazolecarboxylate 95+%
Ethyl-2-amino-4-methyl-5-oxazolecarboxylate
Formel: C₇H₁₀N₂O₃
MW: 170,1659 g/mol
Smältpunkt: 215…217 °C
Storage Temperature: Kylskåp
MDL Number: MFCD09028430
CAS nummer: 79221-15-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR