1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate 95%
Leverantör: Apollo Scientific
|
Varning
|
Synonyms:
1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylic acid ethyl ester, Ethyl 1-Boc-4-oxo-3-piperidinecarboxylate, Ethyl 1-tert-Butoxycarbonyl-4-oxo-3-piperidinecarboxylate, 1-Boc-3-ethoxycarbonyl-4-piperidone, 1-tert-Butyl 3-ethyl 4-oxopiperidine-1,3-dicarboxylate, 1-Boc-4-oxo-3-piperidinecarboxylic acid ethyl ester, 1-tert-Butoxycarbonyl-3-ethoxycarbonyl-4-piperidone
OR40500-10G
OR40500-25G
OR40500-5G
APOSOR40500-10GEA
790
SEK
APOSOR40500-10G
APOSOR40500-25G
APOSOR40500-5G
1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate 95%
1-tert-Butyl-3-ethyl-4-oxopiperidine-1,3-dicarboxylate
Formel:
C₁₃H₂₁NO₅ MW: 271,3100 g/mol Smältpunkt: 52…54 °C Storage Temperature: Omgivande |
MDL Number:
MFCD01862189 CAS nummer: 98977-34-5 |