2-(4,6-Dimethoxypyrimidin-2-yl)cyclohexanecarboxylic acid 95+%
Leverantör: Apollo Scientific
Varning
OR4047-1G OR4047-250MG
APOSOR4047-1GEA 515 SEK
APOSOR4047-1G APOSOR4047-250MG
2-(4,6-Dimethoxypyrimidin-2-yl)cyclohexanecarboxylic acid 95+%
2-(4,6-Dimethoxypyrimidin-2-yl)cyclohexanecarboxylic acid
Racemic mixture of cis isomers
Formel: C₁₃H₁₈N₂O₄
MW: 266,3 g/mol
Smältpunkt: 120…121 °C
Storage Temperature: Omgivande
MDL Number: MFCD01568097
CAS nummer: 633320-99-7