α-Bromo-p-xylene 98%

Leverantör: Apollo Scientific
Fara

Synonyms: alpha-Bromo-p-xylene, 4-Methylbenzyl bromide, 4-(Bromomethyl)toluene

OR40462-100G OR40462-25G OR40462-500G
APOSOR40462-100GEA 1875 SEK
APOSOR40462-100G APOSOR40462-25G APOSOR40462-500G
α-Bromo-p-xylene 98%
α-Bromo-p-xylene
Formel: C₈H₉Br
MW: 185,06 g/mol
Kokpunkt: 218…220 °C
Smältpunkt: 34…36 °C
Flampunkt: 98 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00000181
CAS nummer: 104-81-4
EINECS: 203-240-6
UN: 3417
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR