2-bromo-5-methylthiazole 97%
Leverantör: Apollo Scientific
Synonyms:
2-Bromo-5-methylthiazole
OR40428-10G
OR40428-5G
APOSOR40428-10GEA
1150
SEK
APOSOR40428-10G
APOSOR40428-5G
2-bromo-5-methylthiazole 97%
2-bromo-5-methylthiazole
Formel:
C₄H₄BrNS MW: 178,047 g/mol Kokpunkt: 82…84 °C/9mmHg Storage Temperature: Kylskåp |
MDL Number:
MFCD08460610 CAS nummer: 41731-23-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...