Ethyl-7-bromoimidazo[1,2-a]pyridine-2-carboxylate
Leverantör: Apollo Scientific
Synonyms: Ethyl 7-bromoimidazo[1,2-a]pyridine-2-carboxylate
OR40424-1G OR40424-5G OR40424-25G
APOSOR40424-1GEA 1050 SEK
APOSOR40424-1G APOSOR40424-5G APOSOR40424-25G
Ethyl-7-bromoimidazo[1,2-a]pyridine-2-carboxylate
Ethyl-7-bromoimidazo[1,2-a]pyridine-2-carboxylate
Formel: C₁₀H₉BrN₂O₂
MW: 269,10 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD11505036
CAS nummer: 1187236-18-5