1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-(1,5-Dimethyl-1H-pyrazol-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1,5-Dimethyl-1H-pyrazole-4-boronic acid Pinacol ester
OR40420-1G OR40420-250MG
APOSOR40420-1GEA 670 SEK
APOSOR40420-1G APOSOR40420-250MG
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester 98%
1,5-Dimethyl-1H-pyrazole-4-boronic acid pinacol ester
Formel: C₁₁H₁₉BN₂O₂
MW: 222,0918 g/mol
Smältpunkt: 80…87 °C
Storage Temperature: Frys
MDL Number: MFCD09864189
CAS nummer: 1036991-40-8