[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
Leverantör: Apollo Scientific
Varning
Synonyms: Dichloro 1,1'-bis(diphenylphosphino)ferrocene palladium (II) dichloromethane, [1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) Dichloride Dichloromethane Adduct (1:1), Dichloro[1,1'-bis(diphenylphosphino)ferrocene]palladium(II), complex with dichloromethane, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane adduct, 1,1'-Bis(diphenylphosphino)ferrocene-palladium(II)dichloride dichloromethane complex
OR40417-25G OR40417-1G OR40417-5G
APOSOR40417-25GEA 7175 SEK
APOSOR40417-25G APOSOR40417-1G APOSOR40417-5G
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
[1,1'-Bis(diphenylphosphino)ferrocene]palladium(II) chloride dichloromethane complex
Formel: C₃₅H₃₀Cl₄FeP₂Pd
MW: 816,64 g/mol
Smältpunkt: 275…280 °C
Storage Temperature: Omgivande
MDL Number: MFCD00792899
CAS nummer: 95464-05-4