Phenylphosphonic dichloride
Leverantör: Apollo Scientific
|
Fara
|
Synonyms:
Phenyldichlorophosphine oxide, Dichlorophenylphosphine oxide
OR40402-100G
OR40402-500G
APOSOR40402-100GEA
775
SEK
APOSOR40402-100G
APOSOR40402-500G
Phenylphosphonic dichloride
Phenylphosphonic dichloride
Formel:
C₆H₅Cl₂OP MW: 194,98 g/mol Kokpunkt: 258 °C Smältpunkt: 3 °C Densitet: 1,375 g/cm³ (25 °C) Flampunkt: 204 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00002070 CAS nummer: 824-72-6 EINECS: 212-534-3 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...