4-Amino-3-methylbenzenesulphonic acid

Leverantör: Apollo Scientific
Varning

OR40300-100G OR40300-250G
APOSOR40300-100GEA 465 SEK
APOSOR40300-100G APOSOR40300-250G
4-Amino-3-methylbenzenesulphonic acid
4-Amino-3-methylbenzenesulphonic acid
Formel: C₇H₉NO₃S
MW: 187,22 g/mol
Smältpunkt: >300 °C
Storage Temperature: Omgivande
MDL Number: MFCD00025198
CAS nummer: 98-33-9
EINECS: 202-658-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR