5,6,7,8-Tetrahydroimidazo[1,2-a]pyrazine 95%
Leverantör: Apollo Scientific
Varning
OR40278-1G OR40278-250MG OR40278-5G
APOSOR40278-1GEA 1400 SEK
APOSOR40278-1G APOSOR40278-250MG APOSOR40278-5G
5,6,7,8-Tetrahydroimidazo[1,2-a]pyrazine 95%
5,6,7,8-Tetrahydroimidazo[1,2-a]pyrazine
Formel: C₆H₉N₃
MW: 123,16 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD09701264
CAS nummer: 91476-80-1