2-Bromoisophthalaldehyde 98%

Leverantör: Apollo Scientific
Varning

OR40203-10G OR40203-25G OR40203-5G
APOSOR40203-10GEA 705 SEK
APOSOR40203-10G APOSOR40203-25G APOSOR40203-5G
2-Bromoisophthalaldehyde 98%
2-Bromoisophthalaldehyde
Formel: C₈H₅BrO₂
Smältpunkt: 139…144 °C
Storage Temperature: Omgivande
MDL Number: MFCD08461034
CAS nummer: 79839-49-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR