2-Methoxyphenylboronic acid pinacol ester 97%

Leverantör: Apollo Scientific
Varning

Synonyms: 2-Methoxybenzeneboronic acid pinacol ester, 2-Methoxyphenylboronic acid, pinacol ester, 2-(2-Methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

OR4010-25G OR4010-5G
APOSOR4010-25GEA 5825 SEK
APOSOR4010-25G APOSOR4010-5G
2-Methoxyphenylboronic acid pinacol ester 97%
2-Methoxyphenylboronic acid pinacol ester
Formel: C₁₃H₁₉BO₃
MW: 234,10 g/mol
Smältpunkt: 78…80 °C
Storage Temperature: Omgivande
MDL Number: MFCD05863916
CAS nummer: 190788-60-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR