5-Methoxypyrazine-2-carboxylic acid 95%
Leverantör: Apollo Scientific
Varning
OR40080-1G OR40080-5G OR40080-25G
APOSOR40080-1GEA 685 SEK
APOSOR40080-1G APOSOR40080-5G APOSOR40080-25G
5-Methoxypyrazine-2-carboxylic acid 95%
5-Methoxypyrazine-2-carboxylic acid
Formel: C6H6N2O3
MW: 154.13 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10697773
CAS nummer: 40155-42-8