3-Bromo-5-chloroanisole

Leverantör: Apollo Scientific

Synonyms: 1-Bromo-3-chloro-5-methoxybenzene

OR400559-25G OR400559-5G
APOSOR400559-25GEA 1900 SEK
APOSOR400559-25G APOSOR400559-5G
3-Bromo-5-chloroanisole
3-Bromo-5-chloroanisole
Formel: C₇H₆BrClO
MW: 221,48 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD11035921
CAS nummer: 174913-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR