Methyl-2-(bromomethyl)-5-chlorobenzoate
Leverantör: Apollo Scientific
Varning
Synonyms: Methyl 2-(bromomethyl)-5-chlorobenzoate
OR400300-1G OR400300-250MG OR400300-5G
APOSOR400300-1GEA 1025 SEK
APOSOR400300-1G APOSOR400300-250MG APOSOR400300-5G
Methyl-2-(bromomethyl)-5-chlorobenzoate
Methyl-2-(bromomethyl)-5-chlorobenzoate
Formel: C₉H₈BrClO₂
Kokpunkt: 149…152 °C/8mmHg
Storage Temperature: Kylskåp
MDL Number: MFCD06804577
CAS nummer: 668262-52-0