Methyl-2-bromo-4-chlorobenzoate
Leverantör: Apollo Scientific
Synonyms: 2-Bromo-4-chlorobenzoic acid methyl ester, Methyl 2-bromo-4-chlorobenzoate
OR400102-1G OR400102-25G OR400102-5G
APOSOR400102-1GEA 600 SEK
APOSOR400102-1G APOSOR400102-25G APOSOR400102-5G
Methyl-2-bromo-4-chlorobenzoate
Methyl-2-bromo-4-chlorobenzoate
Formel: C₈H₆BrClO₂
MW: 249,4900 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10566822
CAS nummer: 57381-62-1