3-(Isobutylcarbamoyl)phenylboronic acid 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 3-(Isobutylaminocarbonyl)phenylboronic acid
OR3982-1G OR3982-5G OR3982-25G
APOSOR3982-1GEA 685 SEK
APOSOR3982-1G APOSOR3982-5G APOSOR3982-25G
3-(Isobutylcarbamoyl)phenylboronic acid 97%
3-(Isobutylcarbamoyl)phenylboronic acid
Formel: C₁₁H₁₆BNO₃
Smältpunkt: 224…230 °C
Storage Temperature: Kylskåp
MDL Number: MFCD04115692
CAS nummer: 723282-09-5