3-Aminophenylboronic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: Dihydroxy(3-aminophenyl)borane, (3-Aminophenyl)boronic acid, 3-Aminobenzeneboronic acid, (3-Aminophenyl)metaboric acid
OR3933-5G OR3933-1G OR3933-25G
APOSOR3933-5GEA 295 SEK
APOSOR3933-5G APOSOR3933-1G APOSOR3933-25G
3-Aminophenylboronic acid 98%
3-Aminophenylboronic acid
Formel: C₆H₈BNO₂
MW: 136.95 g/mol
Smältpunkt: 138…143 °C
Storage Temperature: Frys
MDL Number: MFCD00007755
CAS nummer: 30418-59-8