Allylboronic acid pinacol ester 97%
Leverantör: Apollo Scientific
Varning
Synonyms: 2-Allyl-4,4,5,5-Tetramethyl-1,3,2-Dioxaborolane
OR3740-5G OR3740-25G OR3740-100G
APOSOR3740-5GEA 260 SEK
APOSOR3740-5G APOSOR3740-25G APOSOR3740-100G
Allylboronic acid pinacol ester 97%
Allylboronic acid pinacol ester
Formel: C₉H₁₇BO₂
MW: 168.05 g/mol
Kokpunkt: 50…53 °C/5mmHg
Densitet: 0.896 g/cm³ (25 °C)
Flampunkt: 46 °C
Storage Temperature: Omgivande
MDL Number: MFCD00013347
CAS nummer: 72824-04-5
UN: 1993
ADR: 3 III