4-Chloro-8-methylquinoline 97%
Leverantör: Apollo Scientific
|
Varning
|
OR3637-100MG
OR3637-250MG
APOSOR3637-100MGEA
260
SEK
APOSOR3637-100MG
APOSOR3637-250MG
4-Chloro-8-methylquinoline 97%
4-Chloro-8-methylquinoline
Formel:
C₁₀H₈ClN MW: 177.63 g/mol Storage Temperature: Omgivande |
MDL Number:
MFCD00272330 CAS nummer: 18436-73-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...