4,4,5,5-Tetramethyl-2-(3,3,5,5-tetramethylcyclohex-1-en-1-yl)-1,3,2-dioxaborolane 97%
Leverantör: Apollo Scientific
Synonyms: 3,3,5,5-Tetramethyl-1-cyclohexene-1-boronic acid, pinacol ester
OR361637-1G OR361637-250MG
APOSOR361637-1GEA 775 SEK
APOSOR361637-1G APOSOR361637-250MG
4,4,5,5-Tetramethyl-2-(3,3,5,5-tetramethylcyclohex-1-en-1-yl)-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-(3,3,5,5-tetramethylcyclohex-1-en-1-yl)-1,3,2-dioxaborolane
Formel: C₁₆H₂₉BO₂
MW: 264,2113 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16659018
CAS nummer: 859217-85-9