2-Propyl-1,3,2-benzodioxaborole (β-propylocatecholborane) 95%
Leverantör: Apollo Scientific
Fara
Synonyms: 1-Propylboronic acid catechol ester
OR360027-1G OR360027-5G
APOSOR360027-1GEA 2050 SEK
APOSOR360027-1G APOSOR360027-5G
2-Propyl-1,3,2-benzodioxaborole (β-propylocatecholborane) 95%
2-Propyl-1,3,2-benzodioxaborole (β-propylocatecholborane)
Formel: C₉H₁₁BO₂
Storage Temperature: Omgivande
MDL Number: MFCD00799577
CAS nummer: 40218-49-3
UN: 3265
ADR: 8,II