2-Bromo-5-methoxypyridine

Leverantör: Apollo Scientific
Varning

OR3574-5G OR3574-1G
APOSOR3574-5GEA 840 SEK
APOSOR3574-5G APOSOR3574-1G
2-Bromo-5-methoxypyridine
2-Bromo-5-methoxypyridine
Formel: C₆H₆BrNO
Storage Temperature: Frys
MDL Number: MFCD07781221
CAS nummer: 105170-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR