4-Bromo-1-naphthoic acid 97+%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Bromo-1-naphthalenecarboxylic acid
OR3541-25G OR3541-5G
APOSOR3541-25GEA 1225 SEK
APOSOR3541-25G APOSOR3541-5G
4-Bromo-1-naphthoic acid 97+%
4-Bromo-1-naphthoic acid
Formel: C₁₁H₇BrO₂
MW: 251,08 g/mol
Smältpunkt: 217…220 °C
Storage Temperature: Omgivande
MDL Number: MFCD00094221
CAS nummer: 16650-55-8