2-(Methylsulphonyl)phenol 98%

Leverantör: Apollo Scientific
Varning

OR3526-1G OR3526-5G
APOSOR3526-1GEA 1475 SEK
APOSOR3526-1G APOSOR3526-5G
2-(Methylsulphonyl)phenol 98%
2-(Methylsulphonyl)phenol
Formel: C₇H₈O₃S
Smältpunkt: 88…90 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00127516
CAS nummer: 27489-33-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR