4-Methoxypicolinic acid 98%
Leverantör: Apollo Scientific
Varning
Synonyms: 4-Methoxypyridine-2-carboxylic acid
OR3515-1G OR3515-5G
APOSOR3515-1GEA 430 SEK
APOSOR3515-1G APOSOR3515-5G
4-Methoxypicolinic acid 98%
4-Methoxypicolinic acid
Formel: C₇H₇NO₃
MW: 153,14 g/mol
Smältpunkt: 207…208 °C
Storage Temperature: Kylskåp
MDL Number: MFCD04115716
CAS nummer: 29082-91-5