Sodium-2-naphthol-7-sulphonate hydrate 98%
Leverantör: Apollo Scientific
Synonyms: 2-Naphthol-7-sulphonic acid Sodium Salt, F acid Sodium Salt, Cassella's acid Sodium Salt, Sodium 2-naphthol-7-sulphonate, 7-Hydroxy-2-naphthalenesulphonic acid Sodium Salt, Sodium 7-Hydroxy-2-naphthalenesulphonate
OR350346-100G OR350346-500G
APOSOR350346-100GEA 770 SEK
APOSOR350346-100G APOSOR350346-500G
Sodium-2-naphthol-7-sulphonate hydrate 98%
Sodium-2-naphthol-7-sulphonate hydrate
Formel: C₁₀H₈O₄S
MW: 246.21 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00021625
CAS nummer: 135-55-7