3,5-Di-tert-butylphenylboronic acid 97%
Leverantör: Apollo Scientific
Synonyms: 3,5-Di-tert-butylbenzeneboronic acid, 3,5-bis(tert-butyl)phenylboronic acid, (3,5-Di-tert-butylphenyl)boronic acid
OR350137-1G OR350137-10G OR350137-5G
APOSOR350137-1GEA 925 SEK
APOSOR350137-1G APOSOR350137-10G APOSOR350137-5G
3,5-Di-tert-butylphenylboronic acid 97%
3,5-Di-tert-butylphenylboronic acid
Formel: C₁₄H₂₃BO₂
MW: 234,15 g/mol
Smältpunkt: 182 °C
Storage Temperature: Omgivande
MDL Number: MFCD07780212
CAS nummer: 197223-39-5