(3-Bromophenoxy)acetonitrile 95+%
Leverantör: Apollo Scientific
OR346670-1G
OR346670-5G
APOSOR346670-1GEA
1200
SEK
APOSOR346670-1G
APOSOR346670-5G
(3-Bromophenoxy)acetonitrile 95+%
(3-Bromophenoxy)acetonitrile
Formel:
C₈H₆BrNO Storage Temperature: Omgivande |
MDL Number:
MFCD09906440 CAS nummer: 951918-24-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...