(2-Bromophenoxy)acetonitrile 95+%

Leverantör: Apollo Scientific

OR346669-1G OR346669-5G
APOSOR346669-1GEA 1125 SEK
APOSOR346669-1G APOSOR346669-5G
(2-Bromophenoxy)acetonitrile 95+%
(2-Bromophenoxy)acetonitrile
Formel: C₈H₆BrNO
MW: 212,05 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00798567
CAS nummer: 90004-90-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR