(2-tert-Butylphenoxy)acetonitrile 95+%

Leverantör: Apollo Scientific

OR346666-1G OR346666-5G
APOSOR346666-1GEA 895 SEK
APOSOR346666-1G APOSOR346666-5G
(2-tert-Butylphenoxy)acetonitrile 95+%
(2-tert-Butylphenoxy)acetonitrile
Formel: C₁₂H₁₅NO
MW: 189,26 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00184706
CAS nummer: 202821-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR