4-(Bromomethyl)-benzofurazan 95+%

Leverantör: Apollo Scientific
Fara

Synonyms: 4-(Bromomethyl)-2,1,3-benzoxadiazole

OR346588-1G OR346588-250MG
APOSOR346588-1GEA 5750 SEK
APOSOR346588-1G APOSOR346588-250MG
4-(Bromomethyl)-benzofurazan 95+%
4-(Bromomethyl)-benzofurazan
Formel: C₇H₅BrN₂O
Storage Temperature: Omgivande
MDL Number: MFCD00515010
CAS nummer: 32863-30-2
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR