4-Chloro-1-methyl-1H-pyrazole
Leverantör: Apollo Scientific
Varning
OR346437-1G OR346437-5G
APOSOR346437-1GEA 775 SEK
APOSOR346437-1G APOSOR346437-5G
4-Chloro-1-methyl-1H-pyrazole
4-Chloro-1-methyl-1H-pyrazole
Formel: C₄H₅ClN₂
MW: 116,5500 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD08275463
CAS nummer: 35852-81-4