(1-Methyl-1H-benzimidazol-2-yl)methanol 95+%

Leverantör: Apollo Scientific

Synonyms: (1-Methyl-1H-benzo[d]imidazol-2-yl)methanol

OR346407-1G OR346407-5G
APOSOR346407-1GEA 1625 SEK
APOSOR346407-1G APOSOR346407-5G
(1-Methyl-1H-benzimidazol-2-yl)methanol 95+%
(1-Methyl-1H-benzimidazol-2-yl)methanol
Formel: C₉H₁₀N₂O
MW: 162,19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00464051
CAS nummer: 7467-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR