1-Benzofuran-2-sulphonyl chloride

Leverantör: Apollo Scientific
Fara

Synonyms: Benzofuran-2-sulphonyl chloride

OR346249-1G OR346249-500MG OR346249-5G
APOSOR346249-1GEA 4125 SEK
APOSOR346249-1G APOSOR346249-500MG APOSOR346249-5G
1-Benzofuran-2-sulphonyl chloride
1-Benzofuran-2-sulphonyl chloride
Formel: C₈H₅ClO₃S
MW: 216,64 g/mol
Smältpunkt: 79…81 °C
Storage Temperature: Omgivande
MDL Number: MFCD04974038
CAS nummer: 17070-58-5
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR