1-(4-Chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Leverantör: Apollo Scientific
OR346165-1G OR346165-500MG
APOSOR346165-1GEA 0 SEK
APOSOR346165-1G APOSOR346165-500MG
1-(4-Chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
1-(4-Chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carboxylic acid
Formel: C₁₂H₁₁ClN₂O₂
MW: 250,69 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09741134
CAS nummer: 113808-87-0