1-(4-Chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
Leverantör: Apollo Scientific
OR346164-1G OR346164-500MG
APOSOR346164-1GEA 3750 SEK
APOSOR346164-1G APOSOR346164-500MG
1-(4-Chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
1-(4-Chlorophenyl)-3,5-dimethyl-1H-pyrazole-4-carbaldehyde
Formel: C₁₂H₁₁ClN₂O
MW: 234,69 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD09929945
CAS nummer: 54605-74-2