[2-(2-Bromobenzyl)thiazol-4-ylacetic acid

Leverantör: Apollo Scientific

OR346078-1G
APOSOR346078-1GEA 0 SEK
APOSOR346078-1G
[2-(2-Bromobenzyl)thiazol-4-ylacetic acid
[2-(2-Bromobenzyl)thiazol-4-ylacetic acid
Formel: C₁₂H₁₀BrNO₂S
MW: 312.19 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD10568321
CAS nummer: 1053656-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR