3-Methyl-1H-pyrazole-4-carboxylic acid 97%
Leverantör: Apollo Scientific
OR345093-10G OR345093-25G OR345093-5G
APOSOR345093-10GEA 1700 SEK
APOSOR345093-10G APOSOR345093-25G APOSOR345093-5G
3-Methyl-1H-pyrazole-4-carboxylic acid 97%
3-Methyl-1H-pyrazole-4-carboxylic acid
Formel: C5H6N2O2
MW: 126,1133 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD00608153
CAS nummer: 40704-11-8