2-Chloro-3-nitrobenzoic acid 99%
Leverantör: Apollo Scientific
Varning
Synonyms: Mesalazine EP Impurity Q
OR345092-25G OR345092-100G OR345092-5G
APOSOR345092-25GEA 1100 SEK
APOSOR345092-25G APOSOR345092-100G APOSOR345092-5G
2-Chloro-3-nitrobenzoic acid 99%
2-Chloro-3-nitrobenzoic acid
Formel: C₇H₄ClNO₄
Smältpunkt: 182…186 °C
Storage Temperature: Omgivande
MDL Number: MFCD00007069
CAS nummer: 3970-35-2