1,3-Benzothiazole-6-carboxylic acid, Technical Grade
Leverantör: Apollo Scientific
Varning
Synonyms: Benzo[d]thiazole-6-carboxylic acid, Benzothiazole-6-carboxylic acid
OR3419-1G OR3419-5G
APOSOR3419-1GEA 430 SEK
APOSOR3419-1G APOSOR3419-5G
1,3-Benzothiazole-6-carboxylic acid, Technical Grade
1,3-Benzothiazole-6-carboxylic acid
Formel: C₈H₅NO₂S
MW: 179.2 g/mol
Smältpunkt: 245…251 °C
Storage Temperature: Kylskåp
MDL Number: MFCD00111651
CAS nummer: 3622-35-3