8-Bromo-1-naphthoic acid 95%
Leverantör: Apollo Scientific
|
Varning
|
OR3377-1G
OR3377-250MG
APOSOR3377-1GEA
1275
SEK
APOSOR3377-1G
APOSOR3377-250MG
8-Bromo-1-naphthoic acid 95%
8-Bromo-1-naphthoic acid
Formel:
C₁₁H₇BrO₂ MW: 251,0761 g/mol Smältpunkt: 169…171 °C Storage Temperature: Omgivande |
MDL Number:
MFCD00029357 CAS nummer: 1729-99-3 EINECS: 217-045-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...