5-Bromo-2-oxo-6-phenyl-1,2-dihydro-3-pyridinecarbonitrile 95%
Leverantör: Apollo Scientific
OR33638-1G OR33638-5G
APOSOR33638-1GEA 2750 SEK
APOSOR33638-1G APOSOR33638-5G
5-Bromo-2-oxo-6-phenyl-1,2-dihydro-3-pyridinecarbonitrile 95%
5-Bromo-2-oxo-6-phenyl-1,2-dihydro-3-pyridinecarbonitrile
Formel: C₁₂H₇BrN₂O
MW: 275,11 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD16622785
CAS nummer: 1228182-71-5