4-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}morpholine 95%
Leverantör: Apollo Scientific
Synonyms: 4-(1H-Pyrrolo[2,3-d]pyrimidin-4-yl)morpholine
OR33594-1G OR33594-5G
APOSOR33594-1GEA 6325 SEK
APOSOR33594-1G APOSOR33594-5G
4-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}morpholine 95%
4-{7H-Pyrrolo[2,3-d]pyrimidin-4-yl}morpholine
Formel: C₁₀H₁₂N₄O
MW: 204.23 g/mol
Storage Temperature: Omgivande
MDL Number: MFCD18904308
CAS nummer: 90870-83-0